APO455833550 Apollo Scientific Ltd

2-Methyl-5-(pyrrolidin-1-ylsulphonyl)benzeneboronic acid

  • MDL number MFCD08056365
  • Cas Number : 871333-01-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126-128
Synonym 1-[(3-Borono-4-methylbenzene)sulphonyl]pyrrolidine, 2-Borono-4-(pyrrolidin-1-ylsulphonyl)toluene
Purity 98
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=32229

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=32229
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart