APO455833552 Apollo Scientific Ltd

4-Chloro-3-cyanobenzeneboronic acid

  • MDL number MFCD08056360
  • Cas Number : 871332-95-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 300-304
Synonym 5-Borono-2-chlorobenzonitrile
Purity 97
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-chloro-3-cyanobenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-chloro-3-cyanobenzeneboronic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart