APO455833553 Apollo Scientific Ltd

2-Amino-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD06203554
  • Cas Number : 40283-46-3
  • Molecular Weight : 144.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 182-184
Synonym 2-Amino-5-carboxy-1,3-thiazole
Purity 95
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-amino-13-thiazole-5-carboxylic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-13-thiazole-5-carboxylic-acid
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart