APO455833570 Apollo Scientific Ltd

2-(Hydroxymethyl)benzeneboronic acid

  • MDL number MFCD02258966
  • Cas Number : 87199-14-2
  • Molecular Weight : 151.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Boronobenzyl alcohol, (2-Boronophenyl)methanol
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-hydroxymethylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-hydroxymethylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart