APO455833571 Apollo Scientific Ltd

6-(Ethylsulphonyl)-2H-1,4-benzoxazin-3(4H)-one

  • MDL number MFCD00798368
  • Cas Number : 874774-43-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-(Ethylsulphonyl)-4H-benzo[b][1,4]oxazin-3-one, 3,4-Dihydro-3-oxo-2H-1,4-benzoxazin-6-yl ethyl sulphone
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14705

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14705
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart