APO455833573 Apollo Scientific Ltd

1-Methyl-1H-2,1-benzothiazin-4(3H)-one 2,2-dioxide

  • MDL number MFCD00067994
  • Cas Number : 7117-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117-123
Synonym 3,4-Dihydro-1-methyl-2,2,4-trioxo-1H-2,1-benzothiazine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16121

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=16121
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart