APO455833578 Apollo Scientific Ltd

4-(5-Oxopyrazolidin-3-yl)benzeneboronic acid

  • MDL number MFCD08235074
  • Cas Number : 874290-64-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 187-189
Synonym 5-(4-Boronophenyl)pyrazolidin-3-one
Purity 96
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=32316

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=32316
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart