APO455833585 Apollo Scientific Ltd

2-Chloro-5-formylbenzeneboronic acid

  • MDL number MFCD06656269
  • Cas Number : 1150114-78-5
  • Molecular Weight : 184.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Borono-4-chlorobenzaldehyde
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-5-formylbenzeneboronic-acid

Safety Information

signalword Danger
pcodes P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-5-formylbenzeneboronic-acid
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart