APO455833586 Apollo Scientific Ltd

2-Chloro-5-(hydroxymethyl)benzeneboronic acid

  • MDL number MFCD06656270
  • Cas Number : 1003042-59-8
  • Molecular Weight : 186.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Borono-4-chlorobenzyl alcohol, (3-Borono-4-chlorophenyl)methanol
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-5-hydroxymethylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-5-hydroxymethylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart