APO455833598 Apollo Scientific Ltd

N'-(3-Nitropyridin-4-yl)acetohydrazide

  • MDL number MFCD00956008
  • Cas Number : 31481-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(N'-Acetylhydrazino)-3-nitropyridine, Acetic acid N'-(3-nitropyridin-4-yl)hydrazide
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=32295

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=32295
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart