APO455833606 Apollo Scientific Ltd

2-(Methylthio)-6-phenylpyrimidin-4-ol

  • MDL number MFCD00124285
  • Cas Number : 56035-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Methysulphanyl)-6-phenylpyrimidin-4-ol, 2-(Methylsulphanyl)-6-phenylpyrimidin-4(1H)-one, 2-(Methylsulphanyl)-6-phenylpyrimidin-4(3H)-one
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14879

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14879
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart