APO455833610 Apollo Scientific Ltd

Methyl 4-bromo-3-methoxybenzoate

  • MDL number MFCD08436018
  • Cas Number : 17100-63-9
  • Molecular Weight : 245.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58-61
Synonym 2-Bromo-5-(methoxycarbonyl)anisole, Methyl 4-bromo-m-anisate
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-4-bromo-3-methoxybenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-4-bromo-3-methoxybenzoate
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart