APO455833611 Apollo Scientific Ltd

4-Bromo-3-methoxybenzoic acid

  • MDL number MFCD08436017
  • Cas Number : 56256-14-5
  • Molecular Weight : 231.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 221-224
Synonym 2-Bromo-5-carboxyanisole, 4-Bromo-m-anisic acid
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-3-methoxybenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-3-methoxybenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart