APO455833615 Apollo Scientific Ltd

1-(tert-Butyldimethylsilyl)-1H-indole-4-boronic acid

  • MDL number MFCD08436007
  • Cas Number : 351457-64-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-135
Purity 98
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=32343

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=32343
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart