APO455833617 Apollo Scientific Ltd

6-Phenyl-3-thio-1,2,4-triazin-5(2H)-one

  • MDL number MFCD00665863
  • Cas Number : 447-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Phenyl-3-sulphanyl-1,2,4-triazin-5(2H)-one, 2,5-Dihydro-5-oxo-6-phenyl-3-sulphanyl-1,2,4-triazine
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=15022

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=15022
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart