APO455833650 Apollo Scientific Ltd

4-[(Pyridin-2-yl)oxy]benzaldehyde

  • MDL number MFCD08435899
  • Cas Number : 194017-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 45-46
Synonym 2-(4-Formylphenoxy)pyridine
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-pyridin-2-yloxybenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-pyridin-2-yloxybenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart