APO455833665 Apollo Scientific Ltd

3-Bromobenzo[b]furan

  • MDL number MFCD01029425
  • Cas Number : 59214-70-9
  • Molecular Weight : 197.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 35-40
Synonym 3-Bromo-1-benzofuran
Purity 97
Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Boiling point 110
Product Link https://store.apolloscientific.co.uk/product/3-bromobenzobfuran

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromobenzobfuran
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart