APO455833668 Apollo Scientific Ltd

1-[4-Methyl-2-(phenylamino)-1,3-thiazol-5-yl]ethan-1-one

  • MDL number MFCD00129318
  • Cas Number : 31609-42-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-Acetyl-4-methyl-2-(phenylamino)-1,3-thiazole, 5-Acetyl-2-anilino-4-methyl-1,3-thiazole
Purity 98
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=12424

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=12424
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart