APO455833673 Apollo Scientific Ltd

3-Chlorothioanisole

  • MDL number MFCD00040127
  • Cas Number : 4867-37-2
  • Molecular Weight : 158.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Chloro-3-(methylsulphanyl)benzene, 3-Chlorophenyl methyl sulphide, 1-Chloro-3-(methylthio)benzene
Purity 97
Density 1.20
Flash Point 106
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 218
Product Link https://store.apolloscientific.co.uk/product/3-chlorothioanisole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-chlorothioanisole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart