APO455833680 Apollo Scientific Ltd

3-(Pyridin-4-yl)benzoic acid

  • MDL number MFCD05864808
  • Cas Number : 4385-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 268
Synonym 4-(3-Carboxyphenyl)pyridine
Purity 95
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=29474

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=29474
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart