APO455833682 Apollo Scientific Ltd

3-(Aminomethyl)benzaldehyde, N-BOC protected

  • MDL number MFCD06659091
  • Cas Number : 170853-04-0
  • Molecular Weight : 235.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 59
Synonym tert-Butyl (3-formylbenzyl)carbamate, 3-Formylbenzylamine, N-BOC protected
Purity 90
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-aminomethylbenzaldehyde-n-boc-protected

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-aminomethylbenzaldehyde-n-boc-protected
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart