APO455833684 Apollo Scientific Ltd

3-Bromo-2-nitrophenol

  • MDL number MFCD08445675
  • Cas Number : 76361-99-4
  • Molecular Weight : 218.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65-67
Synonym 2-Bromo-6-hydroxynitrobenzene
Purity 96
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-2-nitrophenol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-2-nitrophenol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart