APO455833686 Apollo Scientific Ltd

2-Amino-3-bromophenol

  • MDL number MFCD08445676
  • Cas Number : 116435-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-137
Synonym 2-Bromo-6-hydroxyaniline
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-amino-3-bromophenol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-amino-3-bromophenol
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart