APO455833689 Apollo Scientific Ltd

2,6-Dichloronitrobenzene

  • MDL number MFCD00456795
  • Cas Number : 601-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 65-70
Synonym 1,3-Dichloro-2-nitrobenzene
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 130/8mm
Product Link https://store.apolloscientific.co.uk/product/26-dichloronitrobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/26-dichloronitrobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart