APO455833691 Apollo Scientific Ltd

4-(Pyrrolidin-1-yl)benzeneboronic acid, pinacol ester

  • MDL number MFCD08060504
  • Cas Number : 852227-90-8
  • Molecular Weight : 273.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108.5-109.5
Synonym 1-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrrolidine, 2-[4-(Pyrrolidin-1-yl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Purity 95
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 396
Product Link https://store.apolloscientific.co.uk/product/4-pyrrolidin-1-ylbenzeneboronic-acid-pinacol-ester

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/4-pyrrolidin-1-ylbenzeneboronic-acid-pinacol-ester
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart