APO455833697 Apollo Scientific Ltd

4-Hydroxy-3-iodobenzoic acid

  • MDL number MFCD00234263
  • Cas Number : 37470-46-5
  • Molecular Weight : 264.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-175
Synonym 4-Carboxy-2-iodophenol
Purity 97
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-hydroxy-3-iodobenzoic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-hydroxy-3-iodobenzoic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart