APO455833702 Apollo Scientific Ltd

1-(4-Nitrophenyl)propane-1,2-dione

  • MDL number MFCD01882876
  • Cas Number : 6159-25-7
  • Molecular Weight : 193.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 86-88
Iupac Name 1-(4-nitrophenyl)-1,2-propanedione
Inchi Code 1S/C9H7NO4/c1-6(11)9(12)7-2-4-8(5-3-7)10(13)14/h2-5H,1H3
Inchi Key WCQBWJSIZYUMDJ-UHFFFAOYSA-N

Description

Synonym 4-(2-Oxopropanoyl)nitrobenzene
Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 86-88
Product Link https://store.apolloscientific.co.uk/product/1-4-nitrophenylpropane-12-dione

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-4-nitrophenylpropane-12-dione
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart