APO455833703 Apollo Scientific Ltd

2-Chloro-6-methylpyrazine

  • MDL number MFCD00055032
  • Cas Number : 38557-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Chloro-6-methyl-1,4-diazine
Purity 97
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=31580

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=31580
Availability details for
Quantity
  • Availability
  • Add to Cart