APO455833716 Apollo Scientific Ltd

Ethyl 1,6-dihydro-6-oxo-2-phenylpyrimidine-5-carboxylate

  • MDL number MFCD00695462
  • Cas Number : 55613-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-(Ethoxycarbonyl)-2-phenylpyrimidin-4(3H)-one
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/ethyl-16-dihydro-6-oxo-2-phenylpyrimidine-5-carboxylate

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-16-dihydro-6-oxo-2-phenylpyrimidine-5-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart