APO455833721 Apollo Scientific Ltd

2,3-Diphenyl-6-nitroquinoxaline

  • MDL number MFCD00023917
  • Cas Number : 7466-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,3-Diphenyl-6-nitro-1,4-benzodiazine
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16135

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=16135
pictograms GHS05;GHS06
hstatements H301;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart