APO455833732 Apollo Scientific Ltd

Methyl 3-amino-4-hydroxyphenylacetate

  • MDL number MFCD04125316
  • Cas Number : 78587-72-1
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102-105
Synonym 3-Amino-4-hydroxyphenylacetic acid methyl ester, 2-Amino-4-(2-methoxy-2-oxoethyl)phenol
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-3-amino-4-hydroxyphenylacetate

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/methyl-3-amino-4-hydroxyphenylacetate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart