APO455833733 Apollo Scientific Ltd

Imidazo[1,2-a]pyridine-6-amidoxime

  • MDL number MFCD07366550
  • Cas Number : 885950-24-3
  • Molecular Weight : 176.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191-193
Synonym N'-Hydroxyimidazo[1,2-a]pyridine-6-carboximidamide, N'-Hydroxyimidazo[1,2-a]pyridine-6-carboxamidine
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/imidazo12-apyridine-6-amidoxime

Safety Information

signalword Warning
pcodes P280
MSDS https://store.apolloscientific.co.uk/product/imidazo12-apyridine-6-amidoxime
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart