APO455833734 Apollo Scientific Ltd

8-Hydroxyimidazo[1,2-a]pyridine

  • MDL number MFCD06496222
  • Cas Number : 69214-22-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154-157
Synonym Imidazo[1,2-a]pyridin-8-ol
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/8-hydroxyimidazo12-apyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/8-hydroxyimidazo12-apyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart