APO455833740 Apollo Scientific Ltd

4-Iodophenacyl bromide

  • MDL number MFCD01360521
  • Cas Number : 31827-94-8
  • Molecular Weight : 324.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 113-115
Iupac Name 2-bromo-1-(4-iodophenyl)ethanone
Inchi Code 1S/C8H6BrIO/c9-5-8(11)6-1-3-7(10)4-2-6/h1-4H,5H2
Inchi Key FSIBMLJFLPWMTD-UHFFFAOYSA-N

Description

Synonym 2-Bromo-4'-iodoacetophenone, 2-Bromo-1-(4-iodophenyl)ethan-1-one
Purity 97%
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 113-115
Product Link https://store.apolloscientific.co.uk/product/4-iodophenacyl-bromide

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-iodophenacyl-bromide
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart