APO455833743 Apollo Scientific Ltd

4-Chlorophenacylamine hydrochloride

  • MDL number MFCD00193061
  • Cas Number : 5467-71-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 262
Synonym 2-Amino-1-(4-chlorophenyl)ethan-1-one hydrochloride, alpha-Amino-4'-chloroacetophenone hydrochloride
Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/4-chlorophenacylamine-hydrochloride

Safety Information

signalword Warning
pcodes P260;P312;P280
MSDS https://store.apolloscientific.co.uk/product/4-chlorophenacylamine-hydrochloride
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart