APO455833746 Apollo Scientific Ltd

3-Amino-6-bromo-1H-indazole

  • MDL number MFCD05665872
  • Cas Number : 404827-77-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 237-240
Synonym 6-Bromo-1H-indazol-3-amine
Purity 95
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-amino-6-bromo-1h-indazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-6-bromo-1h-indazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart