APO455833751 Apollo Scientific Ltd

2,4-Dimethyl-5-formyl-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD06202342
  • Cas Number : 253870-02-9
  • Molecular Weight : 167.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 283
Synonym 3,5-Dimethyl-2-formyl-1H-pyrrole-4-carboxylic acid, 3-Carboxy-2,4-dimethyl-5-formyl-1H-pyrrole
Purity 97
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/24-dimethyl-5-formyl-1h-pyrrole-3-carboxylic-acid

Safety Information

signalword Warning
pcodes P280;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/24-dimethyl-5-formyl-1h-pyrrole-3-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart