APO455833756 Apollo Scientific Ltd

2-Methoxy-6-methylpyridine

  • MDL number MFCD04112507
  • Cas Number : 63071-03-4
  • Molecular Weight : 123.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Methoxy-2-picoline
Purity 98
Density 1.01
Flash Point 49
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 156
Product Link https://store.apolloscientific.co.uk/product/2-methoxy-6-methylpyridine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-methoxy-6-methylpyridine
pictograms GHS02;GHS07
hstatements H226;H302+H312+H332;H335
Availability details for
Quantity
  • Availability
  • Add to Cart