APO455833762 Apollo Scientific Ltd

3,4-Dimethoxy-2-hydroxymethylpyridine

  • MDL number MFCD06795941
  • Cas Number : 72830-08-1
  • Molecular Weight : 169.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,4-dimethoxy-2-pyridinyl)methanol
Inchi Code 1S/C8H11NO3/c1-11-7-3-4-9-6(5-10)8(7)12-2/h3-4,10H,5H2,1-2H3
Inchi Key BKTHTLOKUUJKDF-UHFFFAOYSA-N

Description

Synonym (3,4-Dimethoxypyridin-2-yl)methanol, 3,4-Dimethylpicolinyl alcohol
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/34-dimethoxy-2-hydroxymethylpyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/34-dimethoxy-2-hydroxymethylpyridine
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart