APO455833772 Apollo Scientific Ltd

4-Iodo-6-methyl-2-(methylthio)pyrimidine

  • MDL number MFCD00269295
  • Cas Number : 16879-42-8
  • Molecular Weight : 266.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Iodo-6-methyl-2-(methylsulphanyl)pyrimidine, 4-Iodo-6-methyl-2-(methylmercapto)-1,3-diazine
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-iodo-6-methyl-2-methylthiopyrimidine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-iodo-6-methyl-2-methylthiopyrimidine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart