APO455833783 Apollo Scientific Ltd

5-Phenyl-1,3-oxazole

  • MDL number MFCD00067074
  • Cas Number : 1006-68-4
  • Molecular Weight : 145.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40-42
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-phenyl-13-oxazole

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-phenyl-13-oxazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart