APO455833793 Apollo Scientific Ltd

2-Chloro-6-fluoroanisole

  • MDL number MFCD01631573
  • Cas Number : 53145-38-3
  • Molecular Weight : 160.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Chloro-3-fluoro-2-methoxybenzene, 2-Chloro-6-fluorophenyl methyl ether
Purity 97
Flash Point 69
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-6-fluoroanisole

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-6-fluoroanisole
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart