APO455833796 Apollo Scientific Ltd

3-Chloro-2-fluorobenzamide

  • MDL number MFCD00042205
  • Cas Number : 104326-94-5
  • Molecular Weight : 173.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-127
Purity 97
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-2-fluorobenzamide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-2-fluorobenzamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart