APO455833808 Apollo Scientific Ltd

2-Chloro-5-fluorobenzyl alcohol

  • MDL number MFCD01631418
  • Cas Number : 261762-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 68-72
Synonym (2-Chloro-5-fluorophenyl)methanol
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-chloro-5-fluorobenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-5-fluorobenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart