APO455833810 Apollo Scientific Ltd

3-Chloro-4-fluorobenzyl alcohol

  • MDL number MFCD01631550
  • Cas Number : 161446-90-8
  • Molecular Weight : 160.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (3-Chloro-4-fluorophenyl)methanol
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-4-fluorobenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-4-fluorobenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart