APO455833814 Apollo Scientific Ltd

2-Bromo-6-fluorotoluene

  • MDL number MFCD00051438
  • Cas Number : 1422-54-4
  • Molecular Weight : 189.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Bromo-3-fluoro-2-methylbenzene
Purity 98
Density 1.53
Flash Point 76
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 180
Product Link https://store.apolloscientific.co.uk/product/2-bromo-6-fluorotoluene

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-6-fluorotoluene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart