APO455833817 Apollo Scientific Ltd

2-Chloro-4-fluorobenzyl bromide

  • MDL number MFCD00236025
  • Cas Number : 45767-66-6
  • Molecular Weight : 223.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 33-39
Synonym 1-(Bromomethyl)-2-chloro-4-fluorobenzene, alpha-Bromo-2-chloro-4-fluorotoluene
Purity 98
Flash Point 91
Physical Form Solid-Crystals
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 97-99/10mm
Product Link https://store.apolloscientific.co.uk/product/2-chloro-4-fluorobenzyl-bromide

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-4-fluorobenzyl-bromide
pictograms GHS05;GHS07
hstatements H302;H314
Availability details for
Quantity
  • Availability
  • Add to Cart