APO455833822 Apollo Scientific Ltd

3-Chloro-4-fluorobenzyl bromide

  • MDL number MFCD01631551
  • Cas Number : 192702-01-5
  • Molecular Weight : 223.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-(Bromomethyl)-2-chloro-1-fluorobenzene, alpha-Bromo-3-chloro-4-fluorotoluene
Purity 97
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 63-65/0.6mm
Product Link https://store.apolloscientific.co.uk/product/3-chloro-4-fluorobenzyl-bromide

Safety Information

signalword Danger
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-4-fluorobenzyl-bromide
pictograms GHS05;GHS07
hstatements H302;H314
Availability details for
Quantity
  • Availability
  • Add to Cart