APO455833823 Apollo Scientific Ltd

5-Chloro-2-fluorobenzyl bromide

  • MDL number MFCD01631437
  • Cas Number : 71916-91-1
  • Molecular Weight : 223.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(Bromomethyl)-4-chloro-1-fluorobenzene, alpha-Bromo-5-chloro-2-fluorotoluene
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 44-45/0.5mm
Product Link https://store.apolloscientific.co.uk/product/5-chloro-2-fluorobenzyl-bromide

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloro-2-fluorobenzyl-bromide
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart