APO455833830 Apollo Scientific Ltd

2-Chloro-6-fluoro-3-methylbenzoic acid

  • MDL number MFCD01631359
  • Cas Number : 32890-89-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116-117
Synonym 3-Carboxy-2-chloro-4-fluorotoluene, 2-Chloro-6-fluoro-m-toluic acid
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/2-chloro-6-fluoro-3-methylbenzoic-acid

Safety Information

signalword Warning
pcodes P261;P280;P271
MSDS https://store.apolloscientific.co.uk/product/2-chloro-6-fluoro-3-methylbenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart